Learn More
Seracare Life Sciences Inc ANTI-HU IGG (G) F(AB )2 RESAP
Supplier:  Seracare Life Sciences Inc 52200400
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Human IgG heavy chains (gamma). Made in Goat and labeled with alkaline phosphatase. ReserveAP conjugates represent the latest advances in KPL's conjugation chemistry and are more sensitive than KPL's standard AP conjugates. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-69
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.