Learn More
BioVendor, LLC Anti-Human Secretagogin (SCGN), Sheep Polyclonal Antibody (0.1 mg)
SDP
Supplier:  BioVendor, LLC RD184120100
Encompass_Preferred
Polyclonal Antibody. Applications. Western blotting, ELISA, Immunohistochemistry. Source of Antigen E. coliHostsSheepPreparationThe antibody was raised in sheep by immunization with the recombinant Human Secretagogin.Amino Acid SequenceRecombinant Human Secretagogin, total 286 AA. MW: 33.3 kDa (calculated), N-terminal His-tag, 10 extra AA (highlighted), The AA sequence is identical to UniProtKB/Swiss-Prot entry O76038.MKHHHHHHASMDSSREPTLGRLDAAGFWQVWRRFDADEKGYIEEKELDAFFLHMLMKLGTDDTVMKANLHKVKQQFMTTQDASKDGRIRMKELAGMFLSEDENFLLLFRRENPLDSSVEFMQIWRKYDADSSGFISAAELRNFLRDLFLHHKKAISEAKLEEYTGTMMKIFDRNKDGRLDLNDLARILALQENFLLQFKMDACSTEERKRDFEKIFAYYDVSKTGALEGPEVDGFVKDMMELVQPSISGVDLDKFREILLRHCDVNKDGKIQKSELALCLGLKINPSpecies ReactivityHuman. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized recombinant Human Secretagogin.Antibody Content0.1 mg
Catalog No. 50-165-6430
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.