Learn More
Cell Sciences Inc Anti-Human APOE Antibody (1D7), 100 ug
Supplier:  Cell Sciences Inc RHB98603A
Encompass
Species reactivity-Human. Host species-Mouse. Isotype-IgG1. Clonality-Monoclonal. Target-Apo-E, Apolipoprotein E, APOE. Concentration-1 mg/ml. Endotoxin level-Please contact with the lab for this information.Purity>95% as determined by SDS-PAGE.PurificationProtein A/G purified from cell culture supernatant.
Catalog No. NC3251676