Learn More
Chondrex Inc Anti-Human Alpha 2 (IV) NC1 Antibody, Clone H22
Supplier:  Chondrex Inc 7071
Encompass_Preferred
Rat IgG2a monoclonal antibody against the human non-collagenous domain 1 (NC1) of the alpha 2 chain of type IV collagen (alpha 2 (IV) NC1), clone H22, unconjugated. Clone H22 recognizes alpha 2 (IV) NC1 (Epitope: DTLKAGLIR). Use for immunohistochemistry* and immunoblotting. Cross-reacts to dog, human, mouse, and rat alpha 2 (IV) NC1. Purified by Protein G chromatography. Dilute the antibody in buffer containing 2% heterologous serum, such as normal goat serum, to prevent non-specific reactions in immunohistochemistry and immunoblotting.
Catalog No. NC1929635
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.