Learn More
STEMCELL Technologies Anti-Human CD22 Antibody, Clone HIB22, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60083
Encompass_Preferred
The HIB22 antibody reacts with CD22 (Siglec-2), a type I transmembrane glycoprotein expressed in the cytoplasm of pro-B and pre-B cells and on the surface of mature and activated B cells and B cell lymphomas, but not on plasma cells. CD22 functions as an inhibitory receptor for B cell receptor signaling and plays roles in modulating the activation threshold of B cells and controlling CD40 signaling. CD22 comprises two isoforms generated by alternative splicing: CD22α (~130 kDa) and CD22β (~140 kDa). Monomeric CD22β is the most abundant isoform but an αβ heterodimer is also expressed. CD22 is a member of the sialoadhesion subgroup within the immunoglobulin superfamily, with an N-terminal V-type Ig domain followed by six C2-type Ig domains. Binding of the HIB22 antibody reportedly requires the presence of the two N-terminal-most domains. Both the cytoplasmic and surface-bound forms of CD22 are recognized by the HIB22 antibody and its binding does not inhibit CD22-mediated cyto-adhesion.
Catalog No. 50-197-5473
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.