Learn More
STEMCELL Technologies Anti-Human CD32 Antibody, Clone FLI8.26, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60135
Encompass_Preferred
Clone FLI8.26 is a mouse monoclonal antibody that reacts with human and rhesus cynomolgus CD32, an ~40 kDa type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, immunomodulation, and platelet aggregation. Verified for use with ELISA, flow cytometry, and interferometry assays, use this antibody in applications requiring the detection of CD32. The FLI8.26 antibody cross-reacts with monocytes, granulocytes, and platelets. Clone FLI8.26 has been verified to bind to FcγRIIa and FcγRIIb, and reportedly binds to FcγRIIc, which shares an identical amino acid sequence to FcγRIIb in the signal peptide, extracellular and transmembrane domains. Clone FLI8.26 inhibits binding of the FcγRIIa-specific clone IV.3 antibody in flow cytometry analyses. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.