Learn More
STEMCELL Technologies Anti-Human CD326 (EpCAM) Antibody, Clone VU-1D9, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60136
Encompass
Clone VU-1D9 is a mouse monoclonal IgG1 antibody that reacts with the human epithelial cell adhesion molecule (EpCAM or CD326), an ~40 kDa type I transmembrane glycoprotein, functioning as a homophilic and Ca++-independent adhesion molecule. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD326 (EpCAM). EpCAM is not expressed on mesothelial cells or mesotheliomas, and so is widely used as a diagnostic marker to distinguish mesothelioma and carcinoma cells, as well as serving as a target for immunotherapeutic treatment of carcinomas. The VU-1D9 antibody has been used to identify and isolate circulating tumor cells. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5712
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.