Learn More
STEMCELL Technologies Anti-Human CD71 (Transferrin Receptor) Antibody, Clone OKT9, PE, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60106PE.1
Encompass_Preferred
Clone OKT9 is a mouse monoclonal antibody that reacts with human CD71 (transferrin receptor), an ~180 - 190 kDa disulfide-bonded homodimer and type 2 transmembrane glycoprotein expressed at high levels on a broad range of actively proliferating cells, as well as reticulocytes, monocytes, macrophages, and marrow stromal cells. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD71. CD71 has been employed as a marker for evaluating erythroid precursors and for assessing disorders such as erythroid leukemia and myelodysplastic syndrome. It has also served as a target for drug delivery. The OKT9 antibody binds an epitope that reportedly overlaps with that of antibody clone CY1G4 but is distinct from the ligand binding site for transferrin. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5596
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.