Learn More
Advanced ImmunoChemical, Inc. ANTI-HUMAN GLYCATED HBA1C MAB
Supplier:  Advanced ImmunoChemical, Inc. 2GHH1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:75C9Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with glycated (in vitro) synthetic peptide corresponding to the N-terminal end of beta-globin.Specificity:Native human HbA1c, does not cross-react with non-glycated hemoglobin.MAb Isotypes:IgG1 for MAb 75C9Applications:HbA1c immunoassaysPurification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566218