Learn More
Advanced ImmunoChemical, Inc. ANTI-HUMAN HEMOGLOBIN HBA10
Supplier: Advanced ImmunoChemical, Inc. 2HH101 MG
            MONOCLONAL ANTIBODIES/MAbs:Hb4, Hb6Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with glycated (in vitro) synthetic peptide corresponding to the N-terminal end of beta-globin.Specificity:Native human HbA10MAb Isotypes:IgG1 for MAbs Hb4 and Hb6Applications:HbA1c immunoassay.Purification:MAb 4F12: Chromatography on protein A Sepharose and gel-filtration, other MAbs: Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.