Learn More
PBL Assay Science Anti-Human Interferon Lambda Receptor 1 (IFNLR) Mouse Monoclonal Antibody, Clone MMHLR-1 (MAb) [50 micrograms (ug)]
Supplier:  PBL Assay Science 218851
Encompass_Preferred
Mouse monoclonal antibody Anti-Human Interferon Lambda Receptor 1, Clone MMHLR-1 is manufactured by PBL Assay Science (Catalog #21885-1) for research use only (RUO) and packaged in 50 micrograms (ug). This MAb binds to extracellular domains of HuIFNLR and neutralizes human IFNLR1. It is suitable for use in neutralization applications.
Catalog No. 50-153-8030
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.