Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgE Isotype Control, Alexa Fluor™ 546, Novus Biologicals™

Human Polyclonal Antibody
Supplier: Novus Biologicals DDXCH07A546100
Description
IgE Polyclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface), Flow (Intracellular), Immunofluorescence.Specifications
IgE | |
Polyclonal | |
Alexa Fluor 546 | |
PBS, 50% glycerol. with No Preservative | |
IGHE | |
Ion exchange chromatography | |
RUO | |
Isotype Control (Primary) | |
Human | |
IgE |
Flow Cytometry, Immunofluorescence | |
0.5mg/mL | |
Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500 | |
IgE, immunoglobulin heavy constant epsilon | |
Human | |
0.1 mg | |
Adaptive Immunity, Immunology | |
3497 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction