Learn More
Rabbit anti-Human IgG (H+L) Cross-Adsorbed, DyLight 350, Polyclonal, Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Supplier: Thermo Scientific SA510109
Description
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
This antibody is cross-adsorbed and exhibits minimum reactivity to mouse and rat.
Specifications
| Human IgG (H+L) Cross Adsorbed, Human IgG (H+L) | |
| Polyclonal | |
| DyLight 350 | |
| Human IgG-heavy and light chain | |
| 500 μg | |
| Secondary | |
| 4° C | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.5mg/mL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.