Learn More
Advanced ImmunoChemical, Inc. ANTI-HUMAN INSULIN MAB
Supplier: Advanced ImmunoChemical, Inc. 1I11 MG
Monoclonal antibodies / Metabolic Syndrome / Monoclonal mouse anti-human insulin. MAbs:C7C9, D4B8, 7F8, 3A6, 8E2, 7F5. Hybridoma clones have been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with purified human insulin.Specificity: Insulin, human; MAb C7C9 reacts with C-terminal pentapeptide of insulin β-chain. MAbs 7F8, D4B8, 3A6 and 8E2 react with different antigenic determinants. Kass for MAb 7F8 is 7.5 x 109M-1; Kass for MAb D4B8 is 1.5 x 108M-1; Kass for MAb 3A6 is 8.0 x 108M-1; Kass for MAb 8E2 is 6.0 x 108M-1; Kass for MAb C7C9 is 1.0 x 109M-1; Kass for MAb 7F5 is 1.0 x 109M-1. These antibodies cross-react with human proinsulin, bovine insulin (30%) and porcine insulin. No cross-reaction with free C-peptide. MAb isotypes: IgG1, kappa for all MAbs applications: Detection of insulin in two-site enzyme immunoassay.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.