Learn More
Advanced ImmunoChemical, Inc. ANTI-HUMAN SAA MAB
Supplier:  Advanced ImmunoChemical, Inc. 2SAAH1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:SAA6, VSA6 ,VSA25Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cellsof Balb/c mice immunized with human recombinant SAA, conjugated with carrier protein.Specificity:Human recombinant SAAMAb Isotypes:IgG1 for MAbs SAA6, VSA6, VSA25Applications:SAA immunoassay. All MAbs recognize SAA in ELISA and Western blotting.Recommended pairs for sandwich immunoassay (capture-detection):VSA25 – VSA31VSA6 – VSA38Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566228