Learn More
STEMCELL Technologies Anti-Human TRA-1-81 Antibody, Clone TRA-1-81, PE, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60065PE.1
Encompass_Preferred
Clone TRA-1-81 is a mouse monoclonal antibody that reacts with human, rat, and rhesus TRA­1­81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines. Verified for use with flow cytometry, immunocytochemistry, immunofluorescence microscopy, and western blotting assays, use this antibody in applications requiring the detection of TRA-1-81. The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5355
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.