Learn More
Sartorius AHC - Anti-Human Fc Capture
Supplier:  Sartorius 185060
Encompass
Anti-Human Fc Capture (AHC) Biosensors - One tray of 96 biosensors for the immobilization of human IgG or other proteins containing the human Fc region for kinetic analysis
Catalog No. NC9832076
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov