Learn More
FUJIFILM IRVINE SCIENTIFIC INC Anti Iba1, Goat, 100 uL
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 01127991
Encompass_Preferred
Iba1 is a calcium-binding protein with a molecular weight of 17,000 specifically expressed in macrophage and microglia. This product is goat polyclonal antibody that specifically recognize Iba1, and is available for a microglial marker.
Catalog No. NC1801858
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.