Learn More
American Research Products Inc Anti-IDH1 R132H Monoclonal Antibody, 500 µl
Supplier:  American Research Products Inc DIAH09
Encompass
Reactivity: Human, Applications: IHC-P, Clone: H09, Conjugate: Unconjugated, Positive: Control Oligodendroglioma, diffuse astrocytoma, Form: Lyophilized, from PBS, 2% BSA, 0.05% NaN3, pH 7.4., Host: Mouse, Immunogen: Synthetic peptide, amino acid sequence CKPIIIGHHAYGD.
Catalog No. NC1642996
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.