Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

isopentenyl-diphosphate delta isomerase 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-751 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-751 Supplier Abnova Corporation Supplier No. H00003422D01P
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human IDI1 protein.

IDI1 encodes a peroxisomally-localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP), which are the substrates for the successive reaction that results in the synthesis of farnesyl diphosphate and, ultimately, cholesterol. It has been shown in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy that there is reduction in IPP isomerase activity. [provided by RefSeq

Sequence: MMPEINTNHLDKQQVQLLAEMCILIDENDNKIGAETKKNCHLNENIEKGLLHRAFSVFLFNTENKLLLQQRSDAKITFPGCFTNTCCSHPLSNPAELEESDALGVRRAAQRRLKAELGIPLEEVPPEEINYLTRIHYKAQSDGIWGEHEIDYILLVRKNVTLNPDPNEIKSYCYVSKEELKELLKKAASGEIKITPWFKIIAATFLFKWWDNLNHLNQFVDHEKIYRM

Specifications

Antigen isopentenyl-diphosphate delta isomerase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human IDI1 protein.
Formulation PBS with no preservative; pH 7.4
Gene IDI1
Gene Accession No. BC057827.1
Gene Alias IPP1/IPPI1
Gene Symbols IDI1
Host Species Rabbit
Immunogen IDI1 (AAH57827.1, 1 a.a. ∼ 228 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3422
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.