Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IFI16 Ab-1 Matched Antibody Pair, Abnova™

Catalog No. 89023483 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-023-483 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-023-483 Supplier Abnova Corporation Supplier No. H00003428AP11
Add to Cart
Add to Cart
Request a Quote

Includes

Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IFI16 (100μg); Detection antibody: mouse monoclonal anti-IFI16, IgG2a κ (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IFI16

Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a κ
Species Reactivity: Hu

ELISA Pair (Recombinant Protein)

Specifications

Antigen IFI16
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IFI16.
Gene IFI16
Gene Alias IFNGIP1/MGC9466/PYHIN2
Gene Symbols IFI16
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3428
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IFI16 (100 μg) 2. Detection antibody: mouse monoclonal anti-IFI16, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG, IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.