Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ Mouse anti-Pig, Biotin, Clone: P2C11, BD

Catalog No. BDB559958 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559958 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559958 Supplier BD Biosciences Supplier No. 559958
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The P2C11 antibody reacts with pig IFN- γprotein. The immunogen used to generate the P2C11 hybridoma was purified recombinant pig IFN- γ.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications ELISA
Classification Monoclonal
Clone P2C11
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified recombinant pig IFN-γ
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.