Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IFNA2 Ab-2 Matched Antibody Pair, Abnova™

Catalog No. 89003629 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-629 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-629 Supplier Abnova Corporation Supplier No. H00003440AP21

Includes

Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IFNA2 (100μg); Detection antibody: mouse polyclonal anti-IFNA2 (40 ul); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

Host Species: Rabbit, Mouse
Cross Reactivity: Human

ELISA Pair

Specifications

Antigen IFNA2 Ab-2
Classification Polyclonal
Conjugate Unconjugated
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Primary or Secondary Primary
Target Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.