Learn More
anti-Ig Isotype Control Cocktail - A , Clone: G155-228, MOPC-21, MOPC-21, BD
Shop All BD Biosciences ProductsDescription
Flow Cytometry
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | G155-228 |
| Conjugate | FITC/APC/PE |
| Purification Method | Unpurified |
| Quantity | 20 Tests |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.