Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ig, PE-CF594, Clone: R35-95, BD

Catalog No. BDB562302 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562302 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562302 Supplier BD Biosciences Supplier No. 562302
Add to Cart
Add to Cart

Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κ
Immunogen: Mouse pooled immunoglobulin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Immunocytology
Clone R35-95
Concentration 0.2mg/mL
Conjugate PE-CF594
Host Species Rat
Immunogen Mouse pooled immunoglobulin
Purification Method Affinity Purified
Quantity 0.1 mg
Content And Storage 4°C
Isotype IgG2a κ
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.