Learn More
Description
The C48-2 antibody is specific for the hapten trinitrophenol (TNP).
Host Species: Mouse
Clone: C48-2
Isotype: IgE, κImmunogen: TNP-keyhole limpet hemocyanin
ELISA, Isotype Control
Specifications
Specifications
| Applications | ELISA |
| Clone | C48-2 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgE κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.