Learn More
Description
The immunogen used to produce the G235-2356 hybridoma was the hapten trinitrophenol conjugated to a protein carrier. The G235-2356 antibody was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Hamster
Clone: G235-2356
Isotype: IgG1, λ1
Immunogen: Trinitrophenol-KLH
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | G235-2356 |
| Concentration | 0.2mg/mL |
| Conjugate | R-PE |
| Host Species | Hamster |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
| Isotype | IgG1 λ1 |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.