Learn More
anti-IgG1,κ, Clone: R3-34, BD
Supplier: BD Biosciences 562868
Description
Following immunization of a rat with mouse immunoglobulin (Ig), the Ig from the R3-34 hybridoma was identified as a non-reactive clone. The R3-34 immunoglobulin was selected as an Ig isotype control following screening for low background staining on a variety of mouse and human cells and tissues.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Host Species: Rat
Clone: R3-34
Isotype: IgG1, κ
Immunogen: Mouse immunoglobulin
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry | |
R3-34 | |
Brilliant Violet 421 | |
Rat | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse immunoglobulin | |
50 μg | |
Isotype Control (Primary) | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.