Learn More
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by immunocytochemistry. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Immunocytochemistry, Immunohistochemistry (Formalin), Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed), Isotype Control
Specifications
Specifications
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed) |
| Clone | R3-34 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.