Learn More
Description
The A19-3 clone reacts with the hapten, trinitrophenol (TNP). TNP is not expressed on human, mouse, or rat cells. The immunoglobulin from clone A19-3 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Hamster
Clone: A19-3
Isotype: IgG1 κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | A19-3 |
| Concentration | 0.2mg/mL |
| Conjugate | Alexa Fluor 488 |
| Host Species | Hamster |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG1 κ |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.