Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1 κ Isotype Control; Clone: R3-34; BD

Supplier:  BD Biosciences 563283

Catalog No. BDB563283


Only null left
Add to Cart

Description

Description

Host Species: Rat
Clone R3-34
Isotype IgG1, κ

IsCtrl , FC

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
R3-34
Brilliant Violet 711
Rat
Affinity Purified
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse immunoglobulin
50 μg
Isotype Control (Primary)
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.