Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG1, lambda Isotype, Clone: A110-1, BD
Supplier: BD Biosciences 563713
Description
Host Species: Rat
Clone: A110-1
Isotype: IgG1, lambda
Species Reactivity: Ms
Immunogen: P-selectin-IgG1 Fusion
Isotype Control, Flow Cytometry

Specifications
Flow Cytometry | |
A110-1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Trinitrophenyl-Keyhole Limpet Hemocyanin | |
50 μg | |
Isotype Control (Primary) | |
IgG1 λ |
Monoclonal | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction