Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Antibody, Mouse Mouse , Isotype Control, MilliporeSigma™

Catalog No. PP100MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PP100MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PP100MI Supplier MilliporeSigma Supplier No. PP100
Add to Cart
Add to Cart

Mouse Isotype Control

Mouse IgG1 Monoclonal Antibody control validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Applications ELISA
Classification Monoclonal
Concentration 1.95mg/mL
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 100 mM Phosphate buffer, pH 7.0. Contains no preservative.
Host Species Mouse
Purification Method Protein A purfied
Quantity 1 mg
Primary or Secondary Primary
Content And Storage Maintain for 1 year at −20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.