Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG1, κ, Mouse, Alexa Fluor™ 647, Clone: MOPC-21, BD
$289.50 - $313.50
Specifications
Clone | MOPC-21 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugate | Alexa Fluor 647 |
Host Species | Mouse |
Target Species | Human |
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
MOPC-21 | |
Alexa Fluor 647 | |
Human | |
Affinity Purified |
Flow Cytometry, Western Blot | |
Mouse | |
IgG1 κ | |
Undiluted at 4°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title