Learn More
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1, κ
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | MOPC-21 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG1 κ |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.