Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgM κ, Mouse , Clone: C48-6, BD

Catalog No. BDB557275 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557275 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557275 Supplier BD Biosciences Supplier No. 557275
Add to Cart
Add to Cart

The C48-6 antibody is specific for keyhole limpet hemocyanin (KLH).

Host Species: Mouse
Clone: C48-6
Isotype: IgM κImmunogen: Trinitrophenol-KLH

ELISA Standard, Flow Cytometry, Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Clone C48-6
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Trinitrophenol-KLH
Purification Method Affinity Purified
Quantity 0.5 mg
Content And Storage Undiluted at 4°C
Isotype IgM κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.