Learn More
Description
The S1-68.1 antibody has unknown specificity.
Host Species: Mouse
Clone: S1-68.1
Isotype: IgG1 λ
ELISA
Specifications
Specifications
| Applications | ELISA |
| Clone | S1-68.1 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG1 λ |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.