Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1, κ, Mouse, Clone: X40, BD

Catalog No. BDB562438 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562438 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562438 Supplier BD Biosciences Supplier No. 562438
Add to Cart
Add to Cart

Isotype Control

The X40 hybridoma was derived from the fusion of mouse Sp2/0-Ag 14 myeloma cells with spleen cells from BALB/c mice immunized with keyhole limpet hemocyanin (KLH). The hybridoma secretes monoclonal antibody that binds specifically to KLH, an antigen not expressed on human cells or human cell lines.The antibody was conjugated to Brilliant Violet 421 and has been developed in collaboration with Sirigen. With an Ex Max of 407-nm and Em Max at 421-nm, Brilliant Violet 421 can be excited by the violet laser and detected in the standard Pacific Blue filter set (eg, 450/50-nm filter). Brilliant Violet 421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Immunogen: Keyhole limpet hemocyanin (KLH)

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Clone X40
Conjugate Brilliant Violet 421
Host Species Mouse
Immunogen Keyhole limpet hemocyanin (KLH)
Purification Method Affinity Purified
Quantity 50 μg
Content And Storage 4°C, protect from prolonged exposure to light, DO NOT FREEZE
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.