Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1 κ Mouse, FITC, Clone: MOPC-21, Isotype Control, BD

Catalog No. BDB555748 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555748 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555748 Supplier BD Biosciences Supplier No. 555748
Add to Cart
Add to Cart

For low background screening on a variety of mouse and human tissues. anti-IgG1 κ Mouse, FITC, Clone: MOPC-21, Isotype Control, BD immunoglobulin is a mouse myeloma protein. This antibody is routinely tested by flow cytometric analysis.

  • Host Species: Mouse
  • Clone: MOPC-21
  • Isotype: IgG1, κ
  • Applications: Flow Cytometry, Isotype Control
  • Other applications were tested at BD Biosciences Pharmingen during antibody development only
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone MOPC-21
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Content And Storage Undiluted at 4°C. Protect from exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.