Learn More
Description
The MOPC-31C antibody has unknown specificity. The transplantable plasmacytoma MOPC-31C was induced by intraperitoneal injection of mineral oils into BALB/c mice. It was adapted to continuous cell culture by alternate passage in animals.
Host Species: Mouse
Clone: MOPC-31C
Isotype: IgG1 κ
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | MOPC-31C |
| Concentration | 0.2mg/mL |
| Conjugate | PE-Cyanine5 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.