Learn More
anti-IgG1 κ, Mouse, PerCP-Cy™5.5, Clone: MOPC-21, Isotype Control, BD
Shop All BD Biosciences ProductsDescription
This reagent has been used as isotype control in immunophenotypic studies with baboon, rhesus, or cynomolgus peripheral blood cells. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Host Species: Mouse
- Clone: MOPC-21
- Isotype: IgG1 κ
- Applications: Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MOPC-21 |
| Conjugate | PerCP-Cyanine5.5 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.