Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1, PE, Clone: X40, BD

Catalog No. BDB349043 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB349043 100 μg
BDB340013 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB349043 Supplier BD Biosciences Supplier No. 349043
Add to Cart
Add to Cart
The Mouse IgG1 antibody, clone X40, is derived from the hybridization of Sp2/0-Ag 14 mouse myeloma cells with spleen cells from BALB/c mice immunized with KLH. The antibodies react specifically with keyhole limpet hemocyanin (KLH), an antigen not expressed on human cells or human cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Classification Monoclonal
Clone X40
Conjugate PE
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Isotype IgG1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.