Learn More
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Rat
Clone: R3-34
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry, Western Blot |
| Clone | R3-34 |
| Concentration | 0.2mg/mL |
| Conjugate | Alexa Fluor 700 |
| Host Species | Rat |
| Immunogen | Mouse Immunoglobulin |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Target Species | Mouse |
| Content And Storage | Undiluted at 4°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.