Learn More
Description
The B81-3 antibody is specific for keyhole limpet hemocyanin. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Isotype Control
Specifications
Specifications
| Applications | Blocking Assay, Functional Assay, Cytotoxicity Assay |
| Clone | B81-3 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Hamster |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.