Learn More
Description
The C76-47 antibody has unknown specificity.
Host Species: Mouse
Clone: C76-47
Isotype: IgG2a, κImmunogen: Tinitrophenol-keyhole limpet hemocyanin
ELISA Standard
Specifications
Specifications
| Applications | ELISA |
| Clone | C76-47 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Trinitrophenol-KLH |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.