Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2a, Clone: H106.771, SouthernBiotech
SDP

Catalog No. p-3342936 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.2 mg
0.5 mg
1.0 mL
Conjugate:
Alkaline Phosphatase (AP)
Biotin
FITC
None
R-PE
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OB1150-01 0.5 mg None
OB1150-08 0.5 mg Biotin
OB1150-04 1.0 mL Alkaline Phosphatase (AP)
OB1150-02 0.5 mg FITC
OB115009L 0.2 mg R-PE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. OB1150-01 Supplier Southern Biotechnology Associates Supplier No. 115001

Enzyme Immunoassay, Immunofluorescence

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen IgG2a
Classification Monoclonal
Conjugate None
Host Species Rat
Quantity 0.5 mg
Primary or Secondary Primary
Target Species Mouse
Isotype IgG1 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.