Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2a,κ, Clone: R35-95, BD

Catalog No. BDB562952 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562952 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562952 Supplier BD Biosciences Supplier No. 562952
Add to Cart
Add to Cart

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a,κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.

Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κ
Immunogen: Mouse Pooled Immunoglobulin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone R35-95
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Pooled Immunoglobulin
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.