Learn More
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues.
Immunocytochemistry, Immunohistochemistry (Formalin), Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed), Isotype Control
Specifications
Specifications
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed) |
| Clone | R35-95 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.