Learn More
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Applications: Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | G155-178 |
| Conjugate | FITC |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.