Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a Antibody, Mouse Mouse , Isotype Control, MilliporeSigma™

Catalog No. PP102MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PP102MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PP102MI Supplier MilliporeSigma Supplier No. PP102
Add to Cart
Add to Cart

Mouse Isotype Control

Mouse IgG2a Monoclonal Antibody control validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a
Applications ELISA
Classification Monoclonal
Concentration 2.2mg/mL
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing no preservatives.
Host Species Mouse
Quantity 1 mg
Primary or Secondary Primary
Content And Storage Maintain at −20°C in undiluted aliquots for up to 12 months. Do not thaw and refreeze.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.